4-(Boc-amino)-2-bromopyridine

Supplier: Matrix Scientific

Synonyms: tert-Butyl (2-bromopyridin-4-yl)carbamate(2-Bromo-pyridin-4-yl)carbamic acidtert-butyl ester

018201-1G
101805-626EA 76.92 USD
101805-626
4-(Boc-amino)-2-bromopyridine
4-(Boc-amino)-2-bromopyridine
Formula: C₁₀H₁₃BrN₂O₂
MW: 273.13 g/mol
MDL Number: MFCD03791266
CAS Number: 433711-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR