3-Benzyloxyaniline ≥97%

Supplier: Matrix Scientific

017113-5G
101805-432EA 77.05 USD
101805-432
3-Benzyloxyaniline ≥97%
3-Benzyloxyaniline

Hazard code(s): Irritant

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
MDL Number: MFCD00007784
CAS Number: 1484-26-0
EINECS: 216-056-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR