Methyl-5-bromonicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromonicotinate

016822-25G
101805-420EA 27.6 USD
101805-420
Methyl-5-bromonicotinate ≥98%
Methyl 5-bromonicotinate

Hazard code(s): Irritant

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00173668
CAS Number: 29681-44-5

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR