2,5-Dichloropyridine ≥98%

Supplier: Matrix Scientific
Warning

016031-25G 016031-100G
101803-938EA 72.31 USD
101803-938 101803-936
2,5-Dichloropyridine ≥98%
2,5-Dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 190…191 °C
Melting Pt: 59…62 °C
MDL Number: MFCD00006239
CAS Number: 16110-09-1
EINECS: 240-278-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 190-191°C
Melting point 59-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR