m-Anisaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Methoxybenzaldehyde

015675-100G
101803-872EA 181.54 USD
101803-872
m-Anisaldehyde ≥97%
m-Anisaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 101…103 °C (10mm)
Density: 1.118 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003361
CAS Number: 591-31-1
EINECS: 209-712-8

Order Now

Specification Test Results

Boiling point 101-103°C/1
Density 1.118
ND20 1.553

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR