(±)-Isonipecotic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Isonipecotic acidPiperidine-4-carboxylic acid(±)-4-Piperidinecarboxylic acid4-Piperidinecarboxylic acid

012674-100G
101803-180EA 44.62 USD
101803-180
(±)-Isonipecotic acid ≥98%
(±)-Isonipecotic acid

Hazard code(s): Irritant

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: >350 °C
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Merck Index: 13,05208

Order Now

Specification Test Results

Melting point 350°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR