Pyridine-3,5-dicarboxylic acid ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 3,5-Pyridinedicarboxylic acidDinicotinic acid

012644-5G
101803-078EA 21.54 USD
101803-078
Pyridine-3,5-dicarboxylic acid ≥95%
Pyridine-3,5-dicarboxylic acid

Hazard code(s): Irritant

Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 325 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006393
CAS Number: 499-81-0
EINECS: 207-893-8

Order Now

Specification Test Results

Melting point 325°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR