1-(3-Fluorobenzyl)piperazin

Supplier: Matrix Scientific

012114-500MG
101801-938EA 29.23 USD
101801-938
1-(3-Fluorobenzyl)piperazin
1-(3-Fluorobenzyl)piperazin
Formula: C₁₁H₁₅FN₂
MW: 194.25 g/mol
MDL Number: MFCD02242870
CAS Number: 55513-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR