5-Quinolinecarboxylic acid

Supplier: Matrix Scientific
Warning

012039-500MG
101801-794EA 26.45 USD
101801-794
5-Quinolinecarboxylic acid
5-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00047608
CAS Number: 7250-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR