2,6-Dibromopyridine ≥98%

Supplier: Matrix Scientific

011277-25G 011277-100G
101800-304EA 20 USD
101800-304 101800-306
2,6-Dibromopyridine ≥98%
2,6-Dibromopyridine

Hazard code(s): Irritant

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Melting point 118-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR