2-Chloro-5-ethylpyrimidine ≥98%
Supplier: Matrix Scientific
011224-5G
011224-25G
101800-164EA
126.15
USD
101800-164
101800-162
2-Chloro-5-ethylpyrimidine ≥98%
2-Chloro-5-ethylpyrimidine
Hazard code(s): Irritant, Avoid Skin Contact
Formula:
C₆H₇ClN₂ Boiling Pt: 223 °C Density: 1.174 g/cm³ |
MDL Number:
MFCD00799503 CAS Number: 111196-81-7 |
Specification Test Results
Boiling point | 223°C |
Density | 1.174 |
ND20 | 1.521 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...