5,6-Dichloronicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5,6-Dichloropyridine-3-carboxylicacid5,6-Dichloro-3-pyridinecarboxylic acid5,6-Dichloropyridine-3-carboxylic acid

011129-25G
101799-950EA 75.38 USD
101799-950
5,6-Dichloronicotinic acid ≥98%
5,6-Dichloronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 166 °C (dec.)
MDL Number: MFCD00075181
CAS Number: 41667-95-2

Order Now

Specification Test Results

Melting point 166°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR