4-Ethoxyphenyl isothiocyanate

Supplier: Matrix Scientific

Synonyms: 4-Ethoxyphenylisothiocyanate1-Ethoxy-4-isothiocyanato-benzene

010027-500MG
101797-730EA 267.69 USD
101797-730
4-Ethoxyphenyl isothiocyanate
4-Ethoxyphenyl isothiocyanate
Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 146 °C (19 mmHg)
Melting Pt: 75…76 °C
MDL Number: MFCD00041096
CAS Number: 3460-49-9
EINECS: 222-410-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR