Isobutylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Methylpropyl)boronic acid

009791-25G
101797-274EA 49.23 USD
101797-274
Isobutylboronic acid ≥97%
Isobutylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₄H₁₁BO₂
MW: 101.94 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00134156
CAS Number: 84110-40-7

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR