4-Cyanophenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile4-Cyanobenzeneboronic acid pinacol ester

009773-5G
101797-230EA 118.45 USD
101797-230
4-Cyanophenylboronic acid pinacol ester ≥97%
4-Cyanophenylboronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Melting Pt: 95…99 °C
MDL Number: MFCD03093897
CAS Number: 171364-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR