5-Bromo-3-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromopyridyl-5-boronic acid3-Bromopyridine-5-boronic acid

009763-1G
101797-196EA 30.77 USD
101797-196
5-Bromo-3-pyridineboronic acid ≥97%
5-Bromo-3-pyridineboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
MDL Number: MFCD02685634
CAS Number: 452972-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR