4-Aminophenylboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Aminobenzeneboronic acid pinacol ester4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

009752-5G
101797-170EA 32.2 USD
101797-170
4-Aminophenylboronic acid pinacol ester ≥97%
4-Aminophenylboronic acid pinacol ester

Hazard code(s): Irritant, Store in Freezer

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
MDL Number: MFCD02093721
CAS Number: 214360-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR