Perfluoro-1-hexene

009109-2.5G
101796-372EA 248.29 USD
101796-372
Perfluoro-1-hexene
Perfluoro-1-hexene
Formula: C₆F₁₂
MW: 300.05 g/mol
MDL Number: MFCD04038339
CAS Number: 755-25-9
EINECS: 212-047-6

Order Now

Specification Test Results

Boiling point 57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR