1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Iodo-1H,1H,1H,2H,3H,3H-perfluorononane

009012-1G 009012-5G
101796-112EA 272.31 USD
101796-112 101796-110
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane ≥97%
1,1,1,2,2,3,3,4,4,5,5,6,6-Tridecafluoro-8-iodononane

Hazard code(s): Irritant

Formula: C₉H₆F₁₃I
MW: 488.03 g/mol
MDL Number: MFCD00156018
CAS Number: 38550-34-4
EINECS: 253-994-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR