(±)-2-Fluoro-α-methylbenzylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(2-Fluorophenyl)ethylamine
008968-2.5G
008968-500MG
101795-968EA
41.54
USD
101795-968
101795-970
(±)-2-Fluoro-α-methylbenzylamine ≥97%
(±)-2-Fluoro-α-methylbenzylamine
Hazard code(s): Irritant, Refrigerate
Formula:
C₈H₁₀FN MW: 139.17 g/mol |
MDL Number:
MFCD04038307 CAS Number: 74788-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...