2-Fluoro-9-fluorenone ≥98%

Supplier: Matrix Scientific

008948-1G
101795-890EA 290.77 USD
101795-890
2-Fluoro-9-fluorenone ≥98%
2-Fluoro-9-fluorenone

Hazard code(s): Irritant

Formula: C₁₃H₇FO
MW: 198.2 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD00001150
CAS Number: 343-01-1

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR