2,6-Difluoro-4-methoxybenzoic acid ≥98%
Supplier: Matrix Scientific
008924-5G
008924-1G
101795-834EA
44.62
USD
101795-834
101795-836
2,6-Difluoro-4-methoxybenzoic acid ≥98%
2,6-Difluoro-4-methoxybenzoic acid
Hazard code(s): Irritant
Formula:
C₈H₆F₂O₃ Melting Pt: 184…185 °C |
MDL Number:
MFCD02949623 CAS Number: 123843-65-2 |
Specification Test Results
Melting point | 184-185°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...