7-Nitroindole ≥98%

Supplier: Matrix Scientific

008772-5G 008772-1G 008772-25G
101795-422EA 32.31 USD
101795-422 101795-424 101795-420
7-Nitroindole ≥98%
7-Nitroindole

Hazard code(s): Irritant-Harmful, Store in Freezer

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR