3-Fluoroanthranilic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Amino-3-fluorobenzoic acid
008762-5G
008762-25G
101795-390EA
246.15
USD
101795-390
101795-388
3-Fluoroanthranilic acid ≥98%
3-Fluoroanthranilic acid
Hazard code(s): Irritant
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Melting Pt: 185…187 °C |
MDL Number:
MFCD01569395 CAS Number: 825-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...