2,3,5-Trifluoroisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3,5-Trifluoropyridine-4-carboxylic acid

008760-1G
101795-380EA 71.3 USD
101795-380
2,3,5-Trifluoroisonicotinic acid ≥98%
2,3,5-Trifluoroisonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₂F₃NO₂
Melting Pt: 108…110 °C
MDL Number: MFCD03840205
CAS Number: 675602-91-2

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR