2,3,6-Trifluoro-4-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

008758-500MG
101795-374EA 192.31 USD
101795-374
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine ≥98%
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆HF₆N
MW: 201.07 g/mol
Boiling Pt: 102 °C
MDL Number: MFCD03840203
CAS Number: 84940-46-5

Order Now

Specification Test Results

Boiling point 102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR