4-(Trifluoromethoxy)benzamidoxime ≥97%

Supplier: Matrix Scientific

008746-1G
101795-352EA 95.38 USD
101795-352
4-(Trifluoromethoxy)benzamidoxime ≥97%
4-(Trifluoromethoxy)benzamidoxime

Hazard code(s): Irritant

Formula: C₈H₇F₃N₂O₂
Melting Pt: 113…115 °C
MDL Number: MFCD00085175
CAS Number: 56935-71-8

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR