2,5-Dibromopyridine ≥98%

Supplier: Matrix Scientific
Warning

008718-25G 008718-100G
101795-274EA 38.46 USD
101795-274 101795-276
2,5-Dibromopyridine ≥98%
2,5-Dibromopyridine

Hazard code(s): Irritant

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 234…235 °C (1013 hPa)
Melting Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6

Order Now

Specification Test Results

Melting point 93-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR