2-Fluoro-6-nitrotoluene ≥98%
Supplier: Matrix Scientific
008675-100G
008675-25G
101795-148EA
21.54
USD
101795-148
101795-150
2-Fluoro-6-nitrotoluene ≥98%
2-Fluoro-6-nitrotoluene
Hazard code(s): Toxic
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 97…98 °C (11mm) Melting Pt: 6…8 °C Density: 1.277 g/cm³ |
MDL Number:
MFCD00007197 CAS Number: 769-10-8 EINECS: 212-203-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 97-98°C/11m |
ND20 | 1.523 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...