3-Phenylpiperidine

Supplier: Matrix Scientific

008512-1G
101794-826EA 113.85 USD
101794-826
3-Phenylpiperidine
3-Phenylpiperidine

Hazard code(s): Irritant

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 263 °C
Melting Pt: 14.5 °C
Density: 0.967 g/cm³
MDL Number: MFCD00804860
CAS Number: 3973-62-4
EINECS: 223-602-7

Order Now

Specification Test Results

Boiling point 121°C/9mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR