2,4,5-Trimethylbenzoic acid ≥98%

Supplier: Matrix Scientific

007644-1G 007644-5G
101793-166EA 218.46 USD
101793-166 101793-168
2,4,5-Trimethylbenzoic acid ≥98%
2,4,5-Trimethylbenzoic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00075863
CAS Number: 528-90-5

Order Now

Specification Test Results

Melting point 150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR