2,5-Diphenyl-p-benzoquinone ≥97%

Supplier: Matrix Scientific

Synonyms: [1,1:4,1-Terphenyl]-2,5-dione2,5-Diphenylquinone

007583-5G
101793-058EA 76.92 USD
101793-058
2,5-Diphenyl-p-benzoquinone ≥97%
2,5-Diphenyl-p-benzoquinone

Hazard code(s): Irritant

Formula: C₁₈H₁₂O₂
MW: 260.29 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00001600
CAS Number: 844-51-9
EINECS: 212-680-8

Order Now

Specification Test Results

Melting point 210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR