1-Bromo-3-methoxypropane ≥98%
Supplier: Matrix Scientific
007519-25G
007519-100G
102550-908EA
105.8
USD
102550-908
101792-976
1-Bromo-3-methoxypropane ≥98%
1-Bromo-3-methoxypropane
Hazard code(s): Irritant
Formula:
C₄H₉BrO Boiling Pt: 132 °C |
MDL Number:
MFCD02258473 CAS Number: 36865-41-5 |
Specification Test Results
Boiling point | 132°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...