Bicyclohexyl ≥98%

Supplier: Matrix Scientific

007512-25G
101792-954EA 72.31 USD
101792-954
Bicyclohexyl ≥98%
Bicyclohexyl

Hazard code(s): Irritant

Formula: C₁₂H₂₂
MW: 166.31 g/mol
Boiling Pt: 226…228 °C
Density: 0.864 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003815
CAS Number: 92-51-3
EINECS: 202-161-4

Order Now

Specification Test Results

Boiling point 226-228°C
Density 0.864
ND20 1.479

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR