4-Phenyl-1,2,3-thiadiazole ≥98%

Supplier: Matrix Scientific

007470-1G
101792-862EA 49.45 USD
101792-862
4-Phenyl-1,2,3-thiadiazole ≥98%
4-Phenyl-1,2,3-thiadiazole

Hazard code(s): Irritant

Formula: C₈H₆N₂S
MW: 162.22 g/mol
MDL Number: MFCD00087232
CAS Number: 25445-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR