2,3,5,6-Tetrafluorobenzaldehyde ≥98%

Warning

007443-5G 007443-25G
101792-778EA 301.94 USD
101792-778 101792-780
2,3,5,6-Tetrafluorobenzaldehyde ≥98%
2,3,5,6-Tetrafluorobenzaldehyde

Hazard code(s): Irritant, Air Sensitive

Formula: C₇H₂F₄O
MW: 178.09 g/mol
Boiling Pt: 178 °C
Density: 1.525 g/cm³
MDL Number: MFCD00167387
CAS Number: 19842-76-3

Order Now

Specification Test Results

Boiling point 178°C
Density 1.525
ND20 1.469

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR