3-Hydroxyphenylboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: (3-Hydroxyphenyl)boronic acid(3-Hydroxyphenyl)metaboric acidDihydroxy(3-hydroxyphenyl)borane3-Hydroxybenzeneboronic acid

007409-25G 007409-5G
101792-686EA 43.08 USD
101792-686 101792-688
3-Hydroxyphenylboronic acid ≥96%
3-Hydroxyphenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 208…213 °C (dec.)
MDL Number: MFCD01074603
CAS Number: 87199-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR