4-Aminoindole ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Indolamine
, 4-Amino-1H-indole
, Indol-4-ylamine
007259-500MG
101792-274EA
26.15
USD
101792-274
4-Aminoindole ≥98%
4-Aminoindole
Hazard code(s): Irritant-Harmful, Refrigerate, Air Sensitive
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 106…108 °C |
MDL Number:
MFCD01076559 CAS Number: 5192-23-4 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...