1-Bromo-3,5-dichlorobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
3,5-Dichlorobromobenzene
007253-100G
101792-258EA
67.69
USD
101792-258
1-Bromo-3,5-dichlorobenzene ≥98%
1-Bromo-3,5-dichlorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrCl₂ MW: 225.9 g/mol Boiling Pt: 232 °C (757mm) Melting Pt: 75…77 °C |
MDL Number:
MFCD00000584 CAS Number: 19752-55-7 EINECS: 243-270-7 |
Specification Test Results
Boiling point | 232°C/757mm |
Melting point | 75-77°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...