6-Fluoronicotinic acid ≥98%

Synonyms: 2-Fluoro-5-pyridinecarboxylic acid

007209-5G
101792-194EA 199.92 USD
101792-194
6-Fluoronicotinic acid ≥98%
6-Fluoronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄FNO₂
Melting Pt: 146…147 °C
MDL Number: MFCD01859863
CAS Number: 403-45-2

Order Now

Specification Test Results

Melting point 146-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR