3-Nitrobenzophenone ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(3-Nitrophenyl)(phenyl)methanone
, m-Nitrobenzophenone
007086-25G
007086-5G
101791-996EA
181.54
USD
101791-996
101791-994
3-Nitrobenzophenone ≥98%
3-Nitrobenzophenone
Hazard code(s): Irritant
Formula:
C₁₃H₉NO₃ MW: 227.22 g/mol Boiling Pt: 136…138 °C (0.005 mmHg) Melting Pt: 92…94 °C Density: 1.367 g/cm³ (0 °C) |
MDL Number:
MFCD00007257 CAS Number: 2243-80-3 EINECS: 218-819-9 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...