2,6-Naphthalenedicarboxylic acid ≥99%

Supplier: Matrix Scientific

Synonyms: 2,6-Naphthalenedicarboxylic AcidNaphthalene-2,6-dicarboxylic acid

007085-5G
101791-992EA 221.54 USD
101791-992
2,6-Naphthalenedicarboxylic acid ≥99%
2,6-Naphthalenedicarboxylic acid

Hazard code(s): Irritant

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: >300 °C
MDL Number: MFCD00004105
CAS Number: 1141-38-4
EINECS: 214-527-0

Order Now

Specification Test Results

Melting point 300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR