2-Acetamido-5-nitropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Acetylamino-5-nitropyridine

007051-1G 007051-5G
101791-906EA 16.92 USD
101791-906 101791-908
2-Acetamido-5-nitropyridine ≥98%
2-Acetamido-5-nitropyridine

Hazard code(s): Irritant

Formula: C₇H₇N₃O₃
MW: 181.15 g/mol
Melting Pt: 197…200 °C
MDL Number: MFCD00833973
CAS Number: 5093-64-1

Order Now

Specification Test Results

Melting point 197-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR