3,5-Dimethylphenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: Dihydroxy(3,5-dimethylphenyl)borane3,5-Dimethylbenzeneboronic acid(3,5-Dimethylphenyl)metaboric acid

006775-25G
101791-082EA 63.08 USD
101791-082
3,5-Dimethylphenylboronic acid ≥97%
3,5-Dimethylphenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD00185689
CAS Number: 172975-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR