Tribromofluoromethane ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: Fluorotribromomethane

006748-25G 006748-5G
101791-004EA 289.23 USD
101791-004 102832-778
Tribromofluoromethane ≥99%
Tribromofluoromethane

Hazard code(s): Ozone Depleter, Marine Pollutant, Irritant

Formula: Br₃CF
MW: 270.72 g/mol
Boiling Pt: 106 °C
Melting Pt: –74.6 °C
Density: 2.765 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000366
CAS Number: 353-54-8
EINECS: 206-535-8

Order Now

Specification Test Results

Boiling point 106°C
Melting point -74.6°C
Density 2.765
ND20 1.524

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR