Benzoylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-phenylpropanenitrile3-Phenyl-3-oxopropanenitrile

006720-5G 006720-25G
101790-960EA 26.15 USD
101790-960 101790-962
Benzoylacetonitrile ≥98%
Benzoylacetonitrile

Hazard code(s): Irritant

Formula: (C₆H₅)C(O)CH₂CN
MW: 145.16 g/mol
Boiling Pt: 159…160 °C (10mm)
Melting Pt: 82…83 °C
MDL Number: MFCD00001942
CAS Number: 614-16-4
EINECS: 210-369-1

Order Now

Specification Test Results

Boiling point 159-160°C/1
Melting point 82-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR