α,α,α-Trifluoro-4-iodotoluene ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Iodo-4-(trifluoromethyl)benzene
, 4-Iodobenzotrifluoride
, α,α,α-Trifluoro-p-iodotoluene
006693-25G
006693-100G
101790-936EA
60
USD
101790-936
101790-938
α,α,α-Trifluoro-4-iodotoluene ≥97%
α,α,α-Trifluoro-4-iodotoluene
Hazard code(s): Irritant, Stored under Argon
Formula:
C₇H₄F₃I MW: 272.01 g/mol Boiling Pt: 185…186 °C (745mm) Melting Pt: –8.33 °C Density: 1.851 g/cm³ |
MDL Number:
MFCD00039398 CAS Number: 455-13-0 EINECS: 207-234-4 |
Specification Test Results
Boiling point | 185-186°C/7 |
Melting point | -8.33°C |
Density | 1.851 |
ND20 | 1.516 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...