2-Ethylpyrazine ≥98%

Supplier: Matrix Scientific

Synonyms: Ethylpyrazine

006685-100G 006685-25G 006685-5G
101790-908EA 61.54 USD
101790-908 101790-910 101790-906
2-Ethylpyrazine ≥98%
2-Ethylpyrazine

Hazard code(s): Irritant

Formula: C₆H₈N₂
Boiling Pt: 152…153 °C
Density: 0.984 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006149
CAS Number: 13925-00-3
EINECS: 237-691-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 152-153°C
Density 0.984
ND20 1.498

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR