3-Iodobenzhydrazide

Supplier: Matrix Scientific

Synonyms: 3-Iodobenzohydrazide3-Iodobenzoic hydrazide3-Iodobenzene-1-carbohydrazide3-Iodobenzoic acid hydrazide

006683-1G 006683-5G
101790-898EA 44.62 USD
101790-898 101790-900
3-Iodobenzhydrazide
3-Iodobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₇IN₂O
MW: 262.05 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00270106
CAS Number: 39115-94-1

Order Now

Specification Test Results

Melting point 151-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR