5-Fluorocytosine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Amino-5-fluoropyrimidin-2(1H)-oneFlucytosine4-Amino-5-fluoro-2-pyrimidinol5-FC

006676-100G
101790-892EA 52.31 USD
101790-892
5-Fluorocytosine ≥98%
5-Fluorocytosine

Hazard code(s): Irritant, Light Sensitive

Formula: C₄H₄FN₃O
MW: 129.09 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 295…297 °C
MDL Number: MFCD00006035
CAS Number: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Order Now

Specification Test Results

Melting point 295-297°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR